Learn More
R&D Systems™ Cultrex Stem Cell Qualified RGF Basement Membrane Extract
Code produit 18769816
Modifier l'affichage
Click to view available options
Quantité:
10 ml
Conditionnement:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18769816 10 ml 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18769816 Fournisseur R&D Systems™ Code fournisseur 343401002
  • ONLINE EXCLUSIVE PRICE
    631.17€
    562.80€ / 10mL
    Économisez 68.37€
    10% de réduction
 Article restreint
Expédition estimée: 03-12-2026
R&D Systems™ Cultrex Stem Cell Qualified Reduced Growth Factor BME provides an effective surface for the attachment and maintenance of human embryonic stem cells and induced pluripotent stem cells, thereby enabling its use for growth promotion or study of stem cell differentiation.
TRUSTED_SUSTAINABILITY
Spécification
Concentration 8-12 mg/mL (by Lowry assay)
Gamme de produits R&D Systems
Quantité 10 ml
Contenu et stockage Store at -20°C in a manual defrost freezer.