Learn More
Cyclizine Lactate, TRC
Code produit 30367459
Change view
Click to view available options
Quantité:
500 mg
5 g
Conditionnement:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30367459 5 g 5g
30367689 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30367459 Fournisseur Toronto Research Chemicals Code fournisseur TRCC9874405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 5897-19-8
Formule moléculaire C21H28N2O3
Poids moléculaire (g/mol) 356.46
Synonyme 2-Hydroxy-propanoic acid compd. with 1-(diphenylmethyl)-4-methylpiperazine, Lactic acid, compd. with 1-(diphenylmethyl)-4-methylpiperazine, Lactic acid, salt with 1-diphenylmethyl-4-methylpiperazine, 1-(Diphenylmethyl)-4-methyl-piperazine mono(2-hydroxypropanoate), 1-(Diphenylmethyl)-4-methyl-piperazine monolactate, Valoid
Nom IUPAC 1-benzhydryl-4-methylpiperazine;2-hydroxypropanoic acid
SMILES CC(O)C(=O)O.CN1CCN(CC1)C(c2ccccc2)c3ccccc3
Spécification
Nom chimique ou matériau Cyclizine Lactate
Quantité 5 g
Formule InChI InChI=1S/C18H22N2.C3H6O3/c1-19-12-14-20(15-13-19)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;1-2(4)3(5)6/h2-11,18H,12-15H2,1H3;2,4H,1H3,(H,5,6)
Poids de la formule 356.21