Learn More
Cyclohexanone-d10, TRC
Product Code. 30365546
Change view
Click to view available options
Quantité:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30365546 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30365546 Supplier Toronto Research Chemicals Supplier No. TRCC987984250MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Molécules organiques de haute pureté et étalons analytiques, distribués stratégiquement dans le monde entier pour favoriser l'innovation et la réussite commerciale.
TRUSTED_SUSTAINABILITY
Specifications
Nom chimique ou matériau Cyclohexanone-d10
CAS 51209-49-5
Formule moléculaire C6 D10 O
Quantité 250 mg
Synonyme Anon-d10, Anone-d10, Hexanon-d10, Hytrol O-d10, NSC 5711-d10, Nadone-d10, Pimelic Ketone-d6, Pimelin Ketone-d10, Sextone-d10, Cyclohexanone-2,2,3,3,4,4,5,5,6,6-d10, Perdeuterocyclohexanone
Formule InChI InChI=1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2/i1D2,2D2,3D2,4D2,5D2
SMILES [2H]C1([2H])C(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
Nom IUPAC 2,2,3,3,4,4,5,5,6,6-decadeuteriociclohexan-1-ona
Poids moléculaire (g/mol) 108.2
Poids de la formule 108.1359