Learn More
R&D Systems™ Cynomolgus Monkey FAP (XP_005573377) VersaClone cDNA
Spécification
Spécification
| Expression | Fibroblast Activation Protein alpha/FAP |
| Numéro d’adhésion | XP_005573377 |
| Identification génétique (Entrez) | 2191 |
| Alias de gène | 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Cynomolgus Monkey |
| Quantité | 10 μg |