Learn More
Invitrogen™ CytKick™ Autosampler Sample Syringe, 1 mL
Code produit 17246336
Modifier l'affichage
Click to view available options
Quantité:
1 ml
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
17246336 1 ml 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17246336 Fournisseur Invitrogen™ Code fournisseur 100054593CST
Expédition estimée: 01-12-2026
This 1-mL sample syringe is used with the fluidics system of the CytKick and CytKick Max autosampler to move sample from the autosampler to the flow cell of the Attune NxT and Attune CytPix flow cytometers.
This 1-mL sample syringe is used with the fluidics system of the CytKick and CytKick Max autosampler to move sample from the autosampler to the flow cell of the Attune NxT and Attune CytPix flow cytometers. Replacement of this syringe is recommended as part of instrument maintenance.
TRUSTED_SUSTAINABILITY
Spécification
Quantité 1 ml
Capacité 1 mL
À utiliser avec (équipement) CytKick and CytKick Max Autosamplers
Type de produit Autosampler Syringe