Learn More
Invitrogen™ CytKick™ Max Autosampler, refurbished
Code produit 30055931
Modifier l'affichage
Click to view available options
Quantité:
1/boîte
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
30055931 1/boîte 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30055931 Fournisseur Invitrogen™ Code fournisseur A42973R
 Article restreint
Expédition estimée: 28-09-2026
Ajouter au panier
Ajouter au panier
The CytKick Max Autosampler Refurbished is for use with the Attune NxT Acoustic Focusing Cytometer. It enables rapid processing of multiple samples by automatically analyzing samples from 96- and 384-well plates (standard and deep), as well as some custom plate types and microfuge tube racks.
This is a refurbished version of the CytKick Max Autosampler, which is for use with the Attune NxT Acoustic Focusing Cytometer. It enables rapid processing of multiple samples by automatically analyzing samples from 96- and 384-well plates (standard and deep), as well as some custom plate types and microfuge tube racks.
TRUSTED_SUSTAINABILITY
Spécification
Gamme de produits CytKick
Quantité 1/boîte
Type de produit Flow Cytometry Analyzer