Learn More
STEMCELL Technologies DAPI (Hydrochloride)
Code produit 17100362
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantité:
10 mg
Conditionnement:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
17100362 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17100362 Fournisseur STEMCELL Technologies Code fournisseur 75004
 Article restreint
Expédition estimée: 24-11-2026
DNA-labeling dye
  • DAPI (4',6-diamidino-2-phenylindole dihydrochloride) is a blue-fluorescent dye that binds to AT-rich regions of double-stranded DNA
  • Binding is accompanied by an ∼20-fold enhancement in fluorescence, which is directly proportional to the amount of DNA present and has an emission maximum at ∼454 nm
  • The complex is stable for several hours at room temperature and over the pH range 4 - 11
  • DAPI can also bind to RNA, evidently through AU-selective intercalation, though the DAPI/RNA complex emits at a longer wavelength (500 nm) and with only an ∼20% increase in quantum yield
  • DAPI has been widely used as a counterstain to detect nuclei in multicolor fluorescence applications, where its blue fluorescence vividly contrasts with red, yellow, or green fluorescent dyes used to stain other structures
Spécification
CAS 28718-90-3