Learn More
Daprodustat, MedChemExpress
Code produit 16467418
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16467418 10 mg
16459897 5 mg
16457396 100 mg
16433447 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16467418 Fournisseur Medchem Express Code fournisseur HY17608
MedChemExpress Daprodustat (GSK1278863) is an orally active hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor being developed for the treatment of anemia associated with chronic kidney disease.
Daprodustat (GSK1278863) is an orally active hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor being developed for the treatment of anemia associated with chronic kidney disease.
Spécification
Nom chimique ou matériau Daprodustat
Forme physique Solid
CAS 960539-70-2
Couleur Blanc
Quantité 10 mg
Notes de qualité de la pureté Research
Formule moléculaire C19H27N3O6
À utiliser avec (application) COVID-19-immunoregulation
Synonyme GSK1278863
Informations sur la solubilité DMSO : 19.5 mg/mL (49.56 mM; Need ultrasonic and warming)
SMILES O=C(O)CNC(C(C(N1C2CCCCC2)=O)C(N(C3CCCCC3)C1=O)=O)=O
Poids moléculaire (g/mol) 393.43
Poids de la formule 393.43
Pourcentage de pureté 98.71%
Qualité Research