Learn More
Deacetamidine Cyano Dabigatran Ethyl Ester, TRC
Code produit 30367347
Change view
Click to view available options
Quantité:
1 g
10 g
Conditionnement:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30367347 10 g 10g
30364980 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30367347 Fournisseur Toronto Research Chemicals Code fournisseur TRCD19565010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 211915-84-3
Formule moléculaire C27 H26 N6 O3
Poids moléculaire (g/mol) 482.53
Synonyme β-Alanine, N-[[2-[[(4-cyanophenyl)amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-, ethyl ester, 1-Methyl-2-[((4-cyanophenyl)amino)methyl]-benzimidazole-5-carboxamide N-(2-pyridyl)-N-(2-(ethoxycarbonyl)ethyl), Deacetamidine Cyano Dabigatran Ethyl Ester
Nom IUPAC ethyl 3-[[2-[(4-cyanoanilino)methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate
SMILES CCOC(=O)CCN(C(=O)c1ccc2c(c1)nc(CNc3ccc(cc3)C#N)n2C)c4ccccn4
Spécification
Nom chimique ou matériau Ethyl 3-[[2-[(4-Cyanoanilino)methyl]-1-methylbenzimidazole-5-carbonyl]-(2-pyridyl)amino]propanoate
Quantité 10 g
Formule InChI InChI=1S/C27H26N6O3/c1-3-36-26(34)13-15-33(24-6-4-5-14-29-24)27(35)20-9-12-23-22(16-20)31-25(32(23)2)18-30-21-10-7-19(17-28)8-11-21/h4-12,14,16,30H,3,13,15,18H2,1-2H3
Poids de la formule 482.2066