Learn More
Dehydro Azelnidipine, TRC
Code produit 30362397
Change view
Click to view available options
Quantité:
25 mg
Conditionnement:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30362397 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30362397 Fournisseur Toronto Research Chemicals Code fournisseur TRCD22924525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Dehydro Azelnidipine
CAS 918659-10-6
Formule moléculaire C33H32N4O6
Quantité 25 mg
Synonyme 2-Amino-6-methyl-4-(3-nitrophenyl)-3-[1-(diphenylmethyl)-3-azetidinyl]-3,5-pyridinedicarboxylic Acid 5-(1-Methylethyl) Ester
Formule InChI InChI=1S/C33H32N4O6/c1-20(2)42-32(38)27-21(3)35-31(34)29(28(27)24-15-10-16-25(17-24)37(40)41)33(39)43-26-18-36(19-26)30(22-11-6-4-7-12-22)23-13-8-5-9-14-23/h4-17,20,26,30H,18-19H2,1-3H3,(H2,34,35)
SMILES CC(C)OC(=O)c1c(C)nc(N)c(C(=O)OC2CN(C2)C(c3ccccc3)c4ccccc4)c1c5cccc(c5)[N+](=O)[O-]
Poids moléculaire (g/mol) 580.63
Poids de la formule 580.2322