Learn More
MedChemExpress Delcasertib
Code produit 16422946
Encompass_Preferred
Change view
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16422946 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16422946 Fournisseur MedChemExpress Code fournisseur HY106262
MedChemExpress Delcasertib (KAI-9803) is a potent and selective δ-protein kinase C (δPKC) inhibitor. Delcasertib (KAI-9803) could ameliorate injury associated with ischemia and reperfusion in animal models of acute myocardial infarction (MI).
Spécification
Pureté 95.34%
CAS 949100-39-4
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) COVID-19-immunoregulation
Poids moléculaire 2880.28 g/mol
Type de produit Peptides
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité DMSO : ≥ 100 mg/mL (34.72 mM)
Synonyme KAI-9803∣ BMS-875944