Learn More
MedChemExpress Delcasertib
Code produit 16476777
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16476777 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16476777 Fournisseur MedChemExpress Code fournisseur HY106262
MedChemExpress Delcasertib (KAI-9803) is a potent and selective δ-protein kinase C (δPKC) inhibitor. Delcasertib (KAI-9803) could ameliorate injury associated with ischemia and reperfusion in animal models of acute myocardial infarction (MI).
Spécification
Pureté 95.34%
CAS 949100-39-4
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) COVID-19-immunoregulation
Poids moléculaire 2880.28 g/mol
Type de produit Peptides
Quantité 5 mg
Conditions d’expédition Room temperature
Solubilité DMSO : ≥ 100 mg/mL (34.72 mM)
Synonyme KAI-9803∣ BMS-875944