Learn More
Corning™ Denatured Collagen Dissolvable Microcarriers
Code produit 15819037 Tous les produits Corning Produits
Modifier l'affichage
Click to view available options
Quantité:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
15819037 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 15819037 Fournisseur Corning™ Code fournisseur 4981
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
Corning Dissolvable Microcarriers are designed to simplify your cell harvest, separation, and concentration processes. The microcarriers are sterile, and denatured collagen coated to enchance cell attachment, maximizing yield and viability. These microcarriers can be quickly dissolved, allowing easy collection of grown cells without the need for microcarrier separation.
  • Denatured Collagen coated for enhanced cell attachment
  • Highest Sterility Assurance Level (SAL) of 10-6
  • Certified nonpyrogenic
  • Optimized for MSC applications
  • Bead size: Fully hydrated average size 200 to 300µm
  • 5000cm2/gram dry weight
TRUSTED_SUSTAINABILITY
Spécification
Surface de culture 5000 cm2/gram dry weight
À utiliser avec (application) Cell therapy, Vaccine production, Cell Scale-up, Cell Banking
Type de produit Denatured Collagen Dissolvable Microcarrier
Matériau Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions; Denatured Collagen coated
Taille des particules (métrique) 200 to 300 μm fully hydrated
Quantité 10 g