Learn More
MedChemExpress Dermorphin
Code produit 16404016
Encompass_Preferred
Change view
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16404016 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16404016 Fournisseur MedChemExpress Code fournisseur HYP0244
MedChemExpress Dermorphin is a natural heptapeptide μ-opioid receptor (MOR) agonist found in amphibian skin. Inhibition of neuropathic pain.
Spécification
Pureté 98.06%
CAS 77614-16-5
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Neuroscience-Neuromodulation
Poids moléculaire 802.87 g/mol
Type de produit Natural Products
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité H2O : 120 mg/mL (149.46 mM∣Need ultrasonic)