Learn More
MedChemExpress Dermorphin
Code produit 16441898
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16441898 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16441898 Fournisseur MedChemExpress Code fournisseur HYP0244
MedChemExpress Dermorphin is a natural heptapeptide μ-opioid receptor (MOR) agonist found in amphibian skin. Inhibition of neuropathic pain.
Spécification
Pureté 98.06%
CAS 77614-16-5
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Neuroscience-Neuromodulation
Poids moléculaire 802.87 g/mol
Type de produit Natural Products
Quantité 5 mg
Conditions d’expédition Room temperature
Solubilité H2O : 120 mg/mL (149.46 mM∣Need ultrasonic)