Learn More
Desonide, TRC
Code produit 30040795
Change view
Click to view available options
Quantité:
10 mg
50 mg
100 mg
Conditionnement:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30040795 50 mg 50mg
30041025 10 mg 10mg
30006821 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30040795 Fournisseur Toronto Research Chemicals Code fournisseur TRCD29694050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 638-94-8
Formule moléculaire C24 H32 O6
Poids moléculaire (g/mol) 416.51
Synonyme Desonide, 16α,17-[1-Methylethylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione, Budesonide Imp. F (EP), Pregna-1,4-diene-3,20-dione, 11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (6CI,7CI,8CI), 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv., 11β,16α,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione 16,17-acetonide, 11β,21-Dihydroxy-16α,17α-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 16α,17α-(Isopropylidenedioxy)prednisolone, 16α-Hydroxyprednisolone 16,17-acetonide, 16α-Hydroxyprednisolone 16α,17α-acetonide, 16α-Hydroxyprednisolone acetonide, Apolar, D 2083, Desonide, Locapred, Prednacinolone, Prednacinolone acetonide, Sterax, Steroderm, Topifug, Tridesilon
Nom IUPAC 16α,17-[1-Methylethylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Spécification
Nom chimique ou matériau Desonide
Notes de qualité de la pureté HPLC
Quantité 50 mg
Formule InChI InChI=1S/C24H32O6/c1-21(2)29-19-10-16-15-6-5-13-9-14(26)7-8-22(13,3)20(15)17(27)11-23(16,4)24(19,30-21)18(28)12-25/h7-9,15-17,19-20,25,27H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,20+,22-,23-,24+/m0/s1
Poids de la formule 416.2199
Pourcentage de pureté >95%