Learn More
Di-4-ANBDQPQ (Mixture of E/Z isomers, ~90%), TRC
Code produit 17979064
Change view
Click to view available options
Quantité:
0.5 mg
2.5 mg
10 mg
Conditionnement:
0.50mg
10mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17979064 10 mg 10mg
17999724 0.5 mg 0.50mg
17988204 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 17979064 Fournisseur Toronto Research Chemicals Code fournisseur TRCD41643310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1005388-96-4
Formule moléculaire C37H49N3 • 2(Br)
Poids moléculaire (g/mol) 535.802799
Synonyme 4-[4-[6-(Dibutylamino)-2-naphthalenyl]-1,3-butadien-1-yl]-1-[3-(trimethylammonio)propyl]quinolinium Bromide, JPW 6003
SMILES C[N+](C)(CCC[N+]1=CC=C(/C=C/C=C\C2=CC3=C(C=C(C=C3)N(CCCC)CCCC)C=C2)C4=C1C=CC=C4)C.[Br-].[Br-]
Spécification
Nom chimique ou matériau Di-4-ANBDQPQ (Mixture of E/Z isomers)
Quantité 10 mg
Formule InChI InChI=1S/C37H49N3.2BrH/c1-6-8-24-38(25-9-7-2)35-22-21-33-29-31(19-20-34(33)30-35)15-10-11-16-32-23-27-39(26-14-28-40(3,4)5)37-18-13-12-17-36(32)37;;/h10-13,15-23,27,29-30H,6-9,14,24-26,28H2,1-5H3;2*1H/q+2;;/p-2