Learn More
Droxidopa, MedChemExpress
Code produit 16486186
Encompass_Preferred
Change view
Click to view available options
Quantité:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16486186 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16486186 Fournisseur Medchem Express Code fournisseur HY13458
MedChemExpress Droxidopa(L-DOPS) is a potent, orally active norepinephrine precursor. Droxidopa increases standing blood pressure, ameliorates symptoms of orthostatic hypotension and improves standing ability. Droxidopa has the potential for the research of neurogenic orthostatic hypotension (nOH) and alternative ADHD (attention deficit hyperactivity disorder).
Droxidopa(L-DOPS) is a potent, orally active norepinephrine precursor. Droxidopa increases standing blood pressure, ameliorates symptoms of orthostatic hypotension and improves standing ability. Droxidopa has the potential for the research of neurogenic orthostatic hypotension (nOH) and alternative ADHD (attention deficit hyperactivity disorder).
Spécification
Nom chimique ou matériau Droxidopa
CAS 23651-95-8
Couleur Off-White
Notes de qualité de la pureté Research
Formule moléculaire C9H11NO5
Quantité 100 mg
Synonyme L-DOPS DOPS SM5688
Informations sur la solubilité DMSO : 1.8 mg/mL (8.44 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=C(O)C=CC([C@@H](O)[C@@H](C(O)=O)N)=C1
Poids moléculaire (g/mol) 213.19
Poids de la formule 213.19
Pourcentage de pureté 98.04%
Qualité Research
Forme physique Solid
À utiliser avec (application) Neuroscience-Neuromodulation