missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD3

Code produit p-200105784
Change view
Click to view available options
Quantité:
2 ml
5 ml
Conditionnement:
2mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30275450 2 ml 2mL
10725693 5 ml 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30275450 Fournisseur Invitrogen™ Code fournisseur 11152D

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Dynabeads CD3 are superparamagnetic beads coupled with an anti-human CD3 antibody that enable easy isolation or depletion of human CD3+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, PBMC, and tissue digests.

Dynabeads CD3 are superparamagnetic beads coupled with an anti-human CD3 antibody that enable easy isolation or depletion of human CD3+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, peripheral blood mononuclear cells (PBMCs), and tissue digests. Advantages of Dynabeads CD3:

  • Fast and efficient isolation of CD3+ T cells from any sample—no columns required
  • High purity, yield, and viability of positively isolated CD3+ T cells for molecular downstream assays
  • Fast and efficient depletion of CD3+ T cells from any sample.

About Dynabeads CD3

Dynabeads CD3 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal antibody specific for the CD3 membrane antigen mainly expressed mainly on human mature T cells. Because of the bead size, Dynabeads CD3 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads CD3 will also afford excellent recovery of high-purity, viable cells for use in downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate CD3+ cells for cell-based applications, or you need to check your samples with flow cytometry, check out our total human T cell product offerings.

Magnetic bead to based separation offers easy handling

Dynabeads CD3 is added to the sample under continuous mixing to optimize the binding of the Dynabeads to the target cells. By placing the sample on a magnet, the bead-bound target cells are separated from the rest of the sample in just 1 to 2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2 to 3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Automation-ready for KingFisher purification systems

The product is ideal for high-throughput workflows using KingFisher systems. A user guide and scripts for KingFisher Flex and Apex workflows are available for download on this page.

Learn more about Dynabeads products

TRUSTED_SUSTAINABILITY

Spécification

Type de cellule Cellules T (population entière)
Contenu et stockage This Product Contains: 2 mL Dynabeads™ CD3 coated with anti-CD3 monoclonal antibody.
Storage Conditions: 2°C to 8°C. Do not freeze.
Couleur Brown
À utiliser avec (application) Cell Depletion, Positive Isolation for Molecular Applications
Matériau Polystyrène magnétique
Type de produit Antibody Coated Bead
Compatibilité à haut débit Yes
Nbre de cellules Processes ∼8x 10^8 cells total
Viabilité de sortie >95%
Gamme de produits Dynabeads
Pureté ou qualité Research Grade
Quantité 2 ml
Type d’échantillon PBMC, sang
Conditions d’expédition Température ambiante
Numéro de cellule du matériau de départ 1 x 10^7 PBMCs per isolation
Espèces cibles Human
Diamètre (métrique) 4.5 μm
Afficher plus Afficher moins

Usage exclusivement réservé à la recherche. Ne pas utiliser pour des procédures de diagnostic.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.