Learn More
STEMCELL Technologies EasySep™ Human Resting CD4+ T Cell Isolation Kit
Description
- The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Spécification
Spécification
| Type de cellule | T Cells, T Cells, CD4+ |
| À utiliser avec (application) | Cell Isolation |
| Quantité | 1 x 109 cellules |