Learn More
STEMCELL Technologies EasySep™ Mouse CD11c Positive Selection Kit II with Spleen Dissociation Medium
Description
- The EasySep™ Mouse CD11c Positive Selection Kit II isolates highly purified CD11c+ cells from splenocytes or other tissues by immunomagnetic positive selection
- Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet
- Unwanted cells are simply poured off, while desired cells remain in the tube
- Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments
- This product replaces the EasySep™ Mouse CD11c Positive Selection Kit (Catalog #18758) for even faster cell isolations and does not result in the labeling of isolated cells with PE
Spécification
Spécification
| Type de cellule | Dendritic Cells, Granulocytes and Subsets, Monocytes, NK Cells |
| À utiliser avec (application) | Cell Isolation |
| Quantité | 2 x 109 cellules |