Learn More
Invitrogen™ Non-esterified Free Fatty Acids (NEFA/FFA) Colorimetric Assay Kit
Description
The Non-esterified Free Fatty Acids (NEFA/FFA) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of NEFA in animal tissue samples. NEFAs are derived from adiopose tissue, where hormone-sensitive lipase decomposes triglycerides to product fatty acids. These acids circulate freely in the body and, when combined with plasma albumin, can be used as an energy sourced for muscles, organs, and the brain. They are also a substrate of fat synthesis, where the concentration of NEFAs is related to metabolism and endocrine function.
In this assay NEFA react with nantokite to form copper soap, which has a specific absorption peak at 715 nm. The content of NEFA can be calculated indirectly by measuring the OD value.
Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results
Spécification
Spécification
| Plage de dosage | 0.15–1.5 mmol/L |
| Sensibilité du dosage | 0.15 mmol/L |
| Contenu et stockage | Store at 2–8°C. |
| Méthode de détection | Colorimétrique |
| Type d’échantillon | Tissue Homogenates |
| À utiliser avec (équipement) | Microplate Reader |
| Temps d’incubation | 3 min. |
| Coefficient de variation inter-dosage | 5.1 |
| Coefficient de variation intra-dosage | 3.3 |
| Quantité | 96 dosages |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.