Learn More
DiscoverX Embelin
Code produit 17302529
Encompass
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
17302529 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17302529 Fournisseur DiscoverX Code fournisseur 921202
Embelin is an agonist for GPR84 and an inhibitor for XIAP. Provided as 10mg of dry powder.
Embelin is a natural cell-permeable benzoquinone inhibitor of the X-linked inhibitor of apoptosis (XIAP) protein, acting as an anti-tumor, anti-inflammatory, and analgesic. Activates caspases, blocks growth, and promotes apoptosis in tumor cells with high levels of XIAP. Also prevents XIAP- and caspase-dependent inflammation, and inhibits IKK, preventing NF-kB activation. Also acts as an agonist for GPR84. Provided as 10mg of dry powder.
Spécification
Quantité 10 mg