Learn More
MedChemExpress Endothelin 1 (swine, human)
Code produit 16462679
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16462679 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16462679 Fournisseur MedChemExpress Code fournisseur HYP0202
MedChemExpress Endothelin 1 (swine, human) is a synthetic peptide with the sequence of human and swine Endothelin 1, which is a potent endogenous vasoconstrictor. Endothelin 1 acts through two types of receptors ETA and ETB.
Spécification
Pureté 95.44%
CAS 117399-94-7
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) COVID-19-immunoregulation
Poids moléculaire 2491.9 g/mol
Type de produit Reference compound
Quantité 1 mg
Conditions d’expédition Room temperature
Solubilité H2O : 1.82 mg/mL (0.73 mM∣ultrasonic and adjust pH to 3 with HCl)