Learn More
Endoxifen hydrochloride, MedChemExpress
Code produit 16437608
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16437608 10 mg
16417526 100 mg
16413627 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 16437608 Fournisseur Medchem Express Code fournisseur HY18719B
MedChemExpress Endoxifen hydrochloride is a key active metabolite of Tamoxifen (TAM) with higher affinity and specificity to estrogen receptor that also inhibits aromatase activity. Endoxifen hydrochloride has the potential for breast cancer study.
Endoxifen hydrochloride is a key active metabolite of Tamoxifen (TAM) with higher affinity and specificity to estrogen receptor that also inhibits aromatase activity. Endoxifen hydrochloride has the potential for breast cancer study.
Spécification
Nom chimique ou matériau Endoxifen hydrochloride
CAS 1197194-41-4
Couleur Marron
Notes de qualité de la pureté Research
Formule moléculaire C25H28ClNO2
Quantité 10 mg
Informations sur la solubilité DMSO : ≥ 35 mg/mL (85.38 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=CC=C(/C(C2=CC=C(OCCNC)C=C2)=C(C3=CC=CC=C3)\CC)C=C1.[H]Cl
Poids moléculaire (g/mol) 409.95
Poids de la formule 409.95
Pourcentage de pureté 98.0%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death