Learn More
Enrofloxacin
Code produit 15442869
Modifier l'affichage
Click to view available options
Quantité:
5 g
25 g
Conditionnement:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
15442869 5 g 5g
15452869 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 15442869 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur J60023.06
 Article restreint
Expédition estimée: 30-11-2026
Ajouter au panier
A plant fungicide and antibacterial agent | CAS: 93106-60-6 | C19H22FN3O3 | 359.40 g/mol
Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.
Solubility
Soluble in chloroform. Slightly soluble in water. Also soluble in dilute KOH
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 93106-60-6
Formule moléculaire C19H22FN3O3
Poids moléculaire (g/mol) 359.40
Numéro MDL MFCD00792463
Clé InChI SPFYMRJSYKOXGV-UHFFFAOYSA-N
Synonyme 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid
CID PubChem 71188
ChEBI CHEBI:35720
Nom IUPAC 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
SMILES CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O
Spécification
Nom chimique ou matériau Enrofloxacin
Type Enrofloxacin
Forme physique Powder
Point de fusion 221°C to 226°C
Couleur Blanc à jaune
Quantité 5 g
Beilstein 5307824
Merck Index 14,3592
Informations sur la solubilité Soluble in chloroform; Slightly soluble in water; Also soluble in dilute KOH
Poids de la formule 359.39