Learn More
Estropipate, TRC
Code produit 18052240
Change view
Click to view available options
Quantité:
10 mg
50 mg
250 mg
Conditionnement:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18052240 10 mg 10mg
18072340 50 mg 50mg
18052440 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 18052240 Fournisseur Toronto Research Chemicals Code fournisseur TRCE66900010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 7280-37-7
Formule moléculaire C18 H22 O5 S . C4 H10 N2
Poids moléculaire (g/mol) 436.56
Synonyme Estra-1,3,5(10)-trien-17-one, 3-(sulfooxy)-, compd. with piperazine (1:1), Estrone, hydrogen sulfate, compd. with piperazine (1:1) (8CI), Piperazine, compd. with 3-(sulfooxy)estra-1,3,5(10)-trien-17-one (1:1) (9CI), Piperazine, compd. with estrone hydrogen sulfate (1:1) (8CI), 3-Sulfatoxyestra-1,3,5(10)-trien-17-one piperazine salt, Estrone sulfate piperazine salt, Estropipate, Harmogen, Ogen, Piperazine estrone sulfate, Sulestrex, Sulestrex piperazine
Nom IUPAC [(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] hydrogen sulfate;piperazine
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(OS(=O)(=O)O)ccc34)[C@@H]1CCC2=O.C5CNCCN5
Spécification
Nom chimique ou matériau Estropipate
Quantité 10 mg
Formule InChI InChI=1S/C18H22O5S.C4H10N2/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;1-2-6-4-3-5-1/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);5-6H,1-4H2/t14-,15-,16+,18+;/m1./s1
Poids de la formule 436.2032