Learn More
Ethyl 4-[[(methylphenylamino)methylene]amino]benzoate, TRC
Code produit 30375097
Change view
Click to view available options
Quantité:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30375097 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30375097 Fournisseur Toronto Research Chemicals Code fournisseur TRCE925558250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Ethyl 4-(((methylphenylamino)methylene)amino)benzoate
CAS 57834-33-0
Formule moléculaire C17H18N2O2
Quantité 250 mg
Formule InChI InChI=1S/C17H18N2O2/c1-3-21-17(20)14-9-11-15(12-10-14)18-13-19(2)16-7-5-4-6-8-16/h4-13H,3H2,1-2H3
SMILES CCOC(=O)c1ccc(cc1)N=CN(C)c2ccccc2
Poids moléculaire (g/mol) 282.34
Poids de la formule 282.1368