Learn More
Flecainide Acetate, TRC
Code produit 30137250
Change view
Click to view available options
Quantité:
100 mg
1 g
Conditionnement:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30137250 100 mg 100mg
30040823 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30137250 Fournisseur Toronto Research Chemicals Code fournisseur TRCF390010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 54143-56-5
Formule moléculaire C17 H20 F6 N2 O3 . C2 H4 O2
Poids moléculaire (g/mol) 474.39
Synonyme Benzamide, N-(2-piperidinylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)-, acetate (1:1), Benzamide, N-(2-piperidinylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)-, monoacetate (9CI), (±)-Flecainide acetate, 2,5-Bis(2,2,2-trifluoroethoxy)-N-(2-piperidylmethyl)benzamide acetate, Almarytm, Apocard, Ecrinal, Flecainide acetate, R 818, R 818 (pharmaceutical), Tambocor, N-[(RS)-(Piperidin-2-ylmethyl)]-2,5-bis(2,2,2-trifluoroethoxy)benzamide acetate
Nom IUPAC acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
SMILES CC(=O)O.FC(F)(F)COc1ccc(OCC(F)(F)F)c(c1)C(=O)NCC2CCCCN2
Spécification
Nom chimique ou matériau Flecainide Acetate
Quantité 100 mg
Formule InChI InChI=1S/C17H20F6N2O3.C2H4O2/c18-16(19,20)9-27-12-4-5-14(28-10-17(21,22)23)13(7-12)15(26)25-8-11-3-1-2-6-24-11;1-2(3)4/h4-5,7,11,24H,1-3,6,8-10H2,(H,25,26);1H3,(H,3,4)
Poids de la formule 474.1589