Learn More
Fluticasone Acetate, TRC
Code produit 30006990
Change view
Click to view available options
Quantité:
10 mg
100 mg
Conditionnement:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30006990 100 mg 100mg
17971745 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30006990 Fournisseur Toronto Research Chemicals Code fournisseur TRCF599450100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 80474-24-4
Formule moléculaire C24 H29 F3 O5 S
Poids moléculaire (g/mol) 486.54
Synonyme Androsta-1,4-diene-17-carbothioic acid, 17-(acetyloxy)-6,9-difluoro-11-hydroxy-16-methyl-3-oxo-, S-(fluoromethyl) ester, (6alpha,11beta,16alpha,17alpha)-, 6alpha,9-difluoro-17-[[(fluoromethyl)sulfanyl]carbonyl]-11beta-hydroxy-16alpha-methyl-3-oxoandrosta-1,4-dien-17alpha-yl acetate, Fluticasone acetate, Fluticasone Propionate Imp. C (EP), CAF (GW694301X), GW 872664X, TAF (GW675713X), 6-des-fluoro 698 (GW836850X), GSK465433A and GSK1322631A, 5-chloro-furoyl 698 (GSK180878A), TA-TAF dimer (GW871519X), unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), fluticasone acetate (CCI 22341), dithio (GR 269949X)
Nom IUPAC [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] acetate
SMILES C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)C)C(=O)SCF
Spécification
Nom chimique ou matériau Fluticasone acetate
Notes de qualité de la pureté HPLC
Quantité 100 mg
Formule InChI InChI=1S/C24H29F3O5S/c1-12-7-15-16-9-18(26)17-8-14(29)5-6-21(17,3)23(16,27)19(30)10-22(15,4)24(12,32-13(2)28)20(31)33-11-25/h5-6,8,12,15-16,18-19,30H,7,9-11H2,1-4H3/t12-,15+,16+,18+,19+,21+,22+,23+,24+/m1/s1
Poids de la formule 486.1688
Pourcentage de pureté >95%