Learn More
MedChemExpress FMRF
Code produit 16494026
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16494026 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16494026 Fournisseur MedChemExpress Code fournisseur HYP0293
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress FMRF is a peptide consisting of 4 amino acid residues.
Spécification
Pureté 99.08%
CAS 74012-06-9
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Poids moléculaire 599.74 g/mol
Type de produit Peptides
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 100 mg/mL (166.74 mM∣Need ultrasonic)