Learn More
Fosphenytoin disodium, MedChemExpress
Click to view available options
Quantité:
5 mg
10 mg
25 mg
50 mg
100 mg
Description
Fosphenytoin sodium is a phenytoin prodrug with similar anticonvulsant properties.
Specifications
Specifications
| Nom chimique ou matériau | Fosphenytoin disodium |
| CAS | 92134-98-0 |
| Couleur | Blanc |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C16H13N2Na2O6P |
| Quantité | 50 mg |
| Informations sur la solubilité | H2O : ≥ 100 mg/mL (246.16 mM) |
| SMILES | O=C1N(COP(O[Na])(O[Na])=O)C(C(C2=CC=CC=C2)(C3=CC=CC=C3)N1)=O |
| Poids moléculaire (g/mol) | 406.24 |
| Poids de la formule | 406.24 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction