Learn More
MedChemExpress Gap 26
Code produit 16409518
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16409518 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16409518 Fournisseur MedChemExpress Code fournisseur HYP1082
MedChemExpress Gap 26 is a connexin mimetic peptide, composed of residue numbers 63-75 of the first extracellular loop of connexin 43 (gap junction blocker), containing the SHVR amino acid motif.
Spécification
Pureté 99.64%
CAS 197250-15-0
Contenu et stockage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
À utiliser avec (application) COVID-19-immunoregulation
Poids moléculaire 1550.78 g/mol
Type de produit Peptides
Quantité 10 mg
Conditions d’expédition Room temperature
Solubilité H2O : 50 mg/mL (32.24 mM∣Need ultrasonic)