Learn More
R&D Systems™ GAPDH Primer
Product Code. 18236213 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18236213 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18236213 Supplier R&D Systems™ Supplier No. NBP171650SS
  • ONLINE EXCLUSIVE PRICE
    139.47€
    102.90€ / 0.2mL
    Économisez 36.57€
    26% de réduction
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C. Avoid freeze/thaw cycles.
Product Line Novus
Quantity 0.2 mL
For Use With (Application) ChIP
Type Primer