Learn More
R&D Systems™ Geldanamycin
Description
Selectively inhibits heat shock protein 90 (Hsp90). Binds to the ATP site of Hsp90 (Kd = 1.2 μM) and inhibits its chaperone activity. Consequently inhibits activities of oncogenic kinases (e.g. src, Raf), p53 and steroid receptors. Demonstrates antiproliferative effect on breast cancer stem-like cells.
Spécification
Spécification
| CAS | 30562-34-6 |
| Plage de pourcentage du dosage | >98% |
| Quantité | 1 mg |
| Solubilité | Soluble to 10 mM in DMSO |
| Gamme de produits | Tocris |
| Formule moléculaire | C29H40N2O9 |
| Poids de la formule | 560.64g/mol |