Learn More
Gentamicin Monoclonal Antibody (GB73.3), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA5-46053
Description
MA1-10725 detects Gentamicin. MA1-10725 has been successfully used in ELISA applications. The MA1-10725 immunogen is Gentamicin, conjugated to KLH.
Gentamicin is a parenterally administered, broad spectrum aminoglycoside antibiotic typically used for moderate to severe gram negative infections. A complex of three different closely related aminoglycoside sulfates, Gentamicins C1, C2, and C1(subA), obtained from Micromonospora purpurea and related species. They are broad-spectrum antibiotics, but may cause ear and kidney damage. They act to inhibit protein synthesis (genetic translation). [National Center for Biotechnology Information. PubChem Compound Database; CID=3467]
Spécification
| Gentamycin | |
| Monoclonal | |
| 1.1 mg/mL | |
| Gentacycol; Garamycin; Gentavet; Gentamicinum; C21H43N5O7 | |
| Gentamicin, conjugated to KLH. | |
| 100 μg | |
| Primary | |
| Antibody | |
| IgG2a |
| ELISA | |
| GB73.3 | |
| Unconjugated | |
| Mouse | |
| Purified | |
| RUO | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.