Learn More
Globe Scientific Accessories for Elan™ Atac™ 8000 Analyzers
Code produit 31922153
Modifier l'affichage
Click to view available options
À utiliser avec (équipement):
Elan Atac 8000 analyzers
Unit Size:
1000 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. À utiliser avec (équipement) Unit Size
31922153 Elan Atac 8000 analyzers 1000 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31922153 Supplier Globe Scientific Supplier No. 5535
Expédition estimée: 13-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Sample cup for use with Elan Atac 8000 analyzers.
  • For use with Elan Atac 8000 analyzers
  • Material: Polystyrene
  • Quantity: 1000/Pk.
Specifications
Type Sample Cup
À utiliser avec (équipement) Elan Atac 8000 analyzers
Quantité 1000/Pk.