Learn More
MedChemExpress GLP-1(7-37) acetate
Code produit 16472619
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16472619 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16472619 Fournisseur MedChemExpress Code fournisseur HYP0055A
MedChemExpress GLP-1(7-37) acetate is an intestinal insulinotropic hormone that augments glucose induced insulin secretion.
Spécification
Pureté 99.27%
CAS 1450806-98-0
Contenu et stockage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
À utiliser avec (application) Metabolism-sugar/lipid metabolism
Poids moléculaire 3415.72 g/mol
Type de produit Peptides
Quantité 1 mg
Conditions d’expédition Room temperature
Solubilité H2O : 50 mg/mL (14.64 mM∣ultrasonic and adjust pH to 1 with HCl)