Learn More
MedChemExpress GLP-1(7-37) acetate
Code produit 16483966
Encompass_Preferred
Change view
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16483966 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16483966 Fournisseur MedChemExpress Code fournisseur HYP0055A
MedChemExpress GLP-1(7-37) acetate is an intestinal insulinotropic hormone that augments glucose induced insulin secretion.
Spécification
Pureté 99.27%
CAS 1450806-98-0
Contenu et stockage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
À utiliser avec (application) Metabolism-sugar/lipid metabolism
Poids moléculaire 3415.72 g/mol
Type de produit Peptides
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité H2O : 50 mg/mL (14.64 mM∣ultrasonic and adjust pH to 1 with HCl)