missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AcroBiosystems GMP Biotinylated Human CD5 Protein, His,AvitagTM
Description
CD5 protein is a type-I transmembrane glycoprotein found on the surface of thymocytes, T lymphocytes, and a subset of B lymphocytes. It serves to mitigate activating signals from the BCR, allowing B-1 cells to be activated only by strong stimuli. Additionally, CD5 is involved in regulating lipid synthesis and inflammatory responses.
Spécification
Spécification
| Numéro d’adhésion | Uniprot |
| Caractérisation | CD5 Structure This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (AvitagTM). The protein has a calculated MW of 42.2 kDa. The protein migrates as 52 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE) due to glycosylation. |
| Format | Powder-GMP |
| Formule | Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with protectants. Contact us for customized product form or formulation. |
| Test du contrôle qualité | <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative |
| Quantité | 500 μg |
| Source | GMP Biotinylated Human CD5 Protein, His,Avitag (GMP-CD5H24) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # ). Predicted N-terminus: Arg 25 Request for sequence |
| Espèces | Human |
| Stérilité | Sterile |
| Contenu et stockage | -20°C to -70°C for 5 years in lyophilized state; -70°C for 12 months under sterile conditions after reconstitution. Upon receipt, store it immediately at -20°C or lower for long term storage. |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?