missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AcroBiosystems GMP Human BMP-4 / BMP2B (improved sequence) Protein
Description
GMP Human BMP-4 (improved sequence) Protein (GMP-BM4H18) is expressed from E. coli cells. It is a variant of BMP-4 to suit cell culture applications better.
Bone Morphogenetic Protein 4 (BMP4) is a member of growth factor of the TGF-beta superfamily that plays essential roles in many developmental processes, including neurogenesis, vascular development, angiogenesis and osteogenesis. BMP-4 Initiates the canonical BMP signaling cascade by associating with type I receptor BMPR1A and type II receptor BMPR2. BMP-4 can acts in concert with PTHLH/PTHRP to stimulate ductal outgrowth during embryonic mammary development and to inhibit hair follicle induction.
Spécification
Spécification
| Numéro d’adhésion | NCBI |
| Caractérisation | This protein carries no tag. The protein has a calculated MW of 13.2 kDa. The protein migrates as 13 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE). |
| Format | Powder-GMP |
| Formule | Lyophilized from 0.22 μm filtered solution in 20 mM Citric acid, pH 2.2 with protectants. Contact us for customized product form or formulation. |
| Test du contrôle qualité | <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 50 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative |
| Quantité | 50 μg |
| Source | GMP Human BMP-4 (improved sequence) Protein (GMP-BM4H18) is expressed from E. coli cells. It is a variant of BMP-4 to suit cell culture applications better. |
| Activité biologique | GMP Human BMP-4 (improved sequence) Protein (Cat. No. GMP-BM4H18) stimulates Human BMP (Luc) HEK293 Reporter Cell. The specific activity of GMP Human BMP-4 (improved sequence) Protein is >1.90 x 10^3 U/mg (QC tested).∣GMP Human BMP-4 (improved sequence) Protein (Cat. No. GMP-BM4H18) could significantly induce iPSC derived HSPC differentiation, with high expression of CD34 and CD45 (HSPC markers) by flow cytometry analysis and the increase of viable numbers of CD34+ CD45+ cells by cell counting. GMP Human BMP-4 (improved sequence) Protein (Cat. No. GMP-BM4H18) has a similar performance with Human BMP-4 (improved sequence) Protein, premium grade (Cat. No. BM4-H5118).∣The Cell based assay shows that GMP Human BMP-4 (improved sequence) Protein (Cat. No. GMP-BM4H18) is stable at 37°C for 7 hours.∣The Cell based assay shows that GMP Human BMP-4 (improved sequence) Protein (Cat. No. GMP-BM4H18) is stable at 4°C for 180 days.∣The Cell based assay shows that GMP Human BMP-4 (improved sequence) Protein (Cat. No. GMP-BM4H18) is stable after freezing and thawing 3 times.∣The Cell based assay shows batch-to-batch consistency between Acro's GMP and PG BMP-4 (improved sequence). |
| Espèces | Human |
| Stérilité | Sterile |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?