Learn More
GSK467, MedChemExpress
Product Code. 16497607
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
16497607 5 mg
16465646 100 mg
16451117 50 mg
16404868 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497607 Supplier Medchem Express Supplier No. HY116761
This item is not returnable. View return policy
MedChemExpress GSK467 is a cell penetrant and selective KDM5B (JARID1B or PLU1) inhibitor with a Ki of 10 nM and an IC50 of 26 nM. GSK467 shows 180-fold selectivity for KDM4C and no measurable inhibitory effects toward KDM6 or other Jumonji family members.
GSK467 is a cell penetrant and selective KDM5B (JARID1B or PLU1) inhibitor with a Ki of 10 nM and an IC50 of 26 nM. GSK467 shows 180-fold selectivity for KDM4C and no measurable inhibitory effects toward KDM6 or other Jumonji family members.
Specifications
Nom chimique ou matériau GSK467
CAS 1628332-52-4
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C17H13N5O2
Quantité 5 mg
Informations sur la solubilité DMSO : 20.83 mg/mL (65.23 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C2=C(NC(OC3=CN(N=C3)CC4=CC=CC=C4)=N1)C=NC=C2
Poids moléculaire (g/mol) 319.32
Poids de la formule 319.32
Pourcentage de pureté 98.14%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death