Learn More
GW274150, MedChemExpress
Code produit 16494988
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16494988 10 mg
16451237 50 mg
16427727 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 16494988 Fournisseur Medchem Express Code fournisseur HY12119
MedChemExpress GW274150 is a potent, selective, orally active and NADPH-dependent inhibitor of human inducible nitric oxide synthase (iNOS) (IC50=2.19 μM; Kd=40 nM) and rat iNOS (ED50=1.15 μM). GW274150 also displays less potency for both humans or rats endothelial NOS (eNOS) and neuronal NOS (nNOS). GW274150 exerts a protective role in an acute model of lung injury inflammation.
GW274150 is a potent, selective, orally active and NADPH-dependent inhibitor of human inducible nitric oxide synthase (iNOS) (IC50=2.19 μM
Spécification
Nom chimique ou matériau GW274150
CAS 210354-22-6
Quantité 10 mg
Notes de qualité de la pureté Research
Formule moléculaire C8H17N3O2S
À utiliser avec (application) COVID-19-immunoregulation
Informations sur la solubilité H2O : ≥ 62 mg/mL (282.72 mM) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES N[C@@H](CCSCCNC(C)=N)C(O)=O
Poids moléculaire (g/mol) 219.3
Poids de la formule 219.3
Qualité Research