Learn More
Thermo Scientific™ Harvest RC, BT500
Code produit 31767516 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Type:
Chromatography Filter
Conditionnement:
6 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type Conditionnement
31767516 Chromatography Filter 6 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 31767516 Fournisseur Thermo Scientific™ Code fournisseur 7100302871
Expédition estimée: 15-12-2026
The Harvest RC Chromatographic Clarifier is a single stage single-use chromatographic clarification solution designed for harvesting and clarification of modern cell cultures. Clarification with this solution offers fewer processing steps, higher yield and lower production time and costs.
  • Simplification of high cell density cell culture fluid clarification unit operations.
  • Remove cells, cell debris and DNA
  • Optimized for PCV high density CHO cell culture (5-8% PCV)
  • Replacement of primary, secondary and guard membrane clarification stages.
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables.
  • Predictable scaling from discovery to manufacturing.
  • Lower total cost of manufacturing compared to centrifugation and depth filtration.
  • Lower consumption of buffer and water compared to depth filtration.
  • Capsules fit into laboratory to manufacturing scale workflows.
Spécification
Jetable Single-use
Type de filtre Filter Capsule
Matériau (membrane) PES
Stérilité Non-sterile
Quantité 6/Cs.
Type Chromatography Filter