Learn More
Brannan Heavy Duty Anemometer with Temperature and Humidity Meter
Code produit 31810744
Change view
Click to view available options
Type de gaz:
Oxygen
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de gaz unitSize
31810744 Oxygen 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 31810744 Fournisseur Brannan Code fournisseur 13/490/0
Anemometer for air velocity and air volume.
  • Measures air velocity and air volume with thermometer and humidity meter
  • Large LCD with green, yellow and red light indication
  • Supplied with calibration certificate and sturdy protective pouch
  • Air velocity level indication
  • Max min / Avg / data hold function
  • Low battery indication
  • Audible alarm
  • Auto power off
Spécification
Type Anemometer/Thermometer
Précision ±3% (Full Scale)
Débit d’air 0.4 to 35 m/sec. (1.4 to 122.5 km/hr.)
Matériau du coffret ABS Plastic
Float Material Polypropylene
Type de gaz Oxygen
Pression nominale 100 psig
Affichage LCD
À utiliser avec (application) Wind Speed Meter
Détails du bloc d’alimentation 3 x AA Batteries