Learn More
Heparan Sulfate, MedChemExpress
Code produit 16456367
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16456367 5 mg
16452806 25 mg
16433638 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 16456367 Fournisseur Medchem Express Code fournisseur HY101916
MedChemExpress Heparan sulfate, a complex and linear polysaccharide, exists as part of glycoproteins named heparan sulfate proteoglycans, which are expressed abundantly on the cell surface and in the extracellular matrix.
Heparan sulfate, a complex and linear polysaccharide, exists as part of glycoproteins named heparan sulfate proteoglycans, which are expressed abundantly on the cell surface and in the extracellular matrix.
Spécification
Nom chimique ou matériau Heparan Sulfate
Forme physique Solid
CAS 9050-30-0
Couleur Blanc
Quantité 5 mg
Notes de qualité de la pureté Research
Formule moléculaire C12H19NO20S3 (monomer)
À utiliser avec (application) Cancer-Kinase/protease
Informations sur la solubilité H2O : 100 mg/mL (Need ultrasonic)
SMILES O=S(OCC1O[C@@H](OC)[C@H](NS(=O)(O)=O)[C@@H](OS(=O)(O)=O)[C@H]1O[C@@H]2OC(C(O)=O)[C@H](O[C@@H]3OC(COS(=O)(O)=O)[C@H](O[C@@H]4OC(C(O)=O)[C@H](OC)[C@H](O)[C@H]4OS(=O)(O)=O)[C@H](O)[C@H]3NS(=O)(O)=O)[C@H](O)[C@H]2O)(O)=O.[n]
Qualité Research