Learn More
MedChemExpress HOE 32021
Code produit 16486806
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16486806 100 mg
16486628 10 mg
16429197 5 mg
16412727 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16486806 Fournisseur MedChemExpress Code fournisseur HY15562
MedChemExpress Hoechst stains are part of a family of blue fluorescent dyes used to stain DNA.
Spécification
Contenu et stockage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Quantité 100 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 50 mg/mL (114.02 mM; Need ultrasonic)
Poids moléculaire 438.52 g/mol
CAS 23623-06-5