Learn More
MedChemExpress Hoechst 33342 analog
Code produit 16486698
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Conditionnement:
100mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16486698 10 mg -
16459257 5 mg -
16416856 100 mg 100mg
16402787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16486698 Fournisseur MedChemExpress Code fournisseur HY15627
MedChemExpress Hoechst 33342 analog is an anglog of Hoechst 33342, which is a DNA minor groove binder used fluorochrome for visualizing cellular DNA.
Spécification
Contenu et stockage Powder -20°C 3 years ∣ In solvent -80°C 6 months, -20°C 1 month
Quantité 10 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 100 mg/mL (169.32 mM; Need ultrasonic)
Poids moléculaire 590.59 g/mol
CAS 178481-68-0