Learn More
R&D Systems™ Human ALDH1A3 (NP_000684.2) VersaClone cDNA
Spécification
Spécification
| Expression | ALDH1A3 |
| Identification génétique (Entrez) | 220 |
| Alias de gène | aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3 |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantité | 10 μg |