missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ARTS Synthetic Peptide
Description
ARTS Synthetic Peptide, PEP-0123, from Invitrogen.
Apoptosis is related to many diseases and development. Mitochondrial proteins, such as cytochrome c, Apaf-1, and AIF play important role in apoptosis. A novel mitochondrial septin-like protein was identified recently and designated ARTS for apoptosis related protein in TGF-beta signaling pathway. ARTS that is encoded by the human septin H5/PNUTL2/CDCrel2b gene is located to mitochondria and translocates to the nucleus when apoptosis occurs. ARTS is expressed in many tissues. It enhances cell death induced by TGF-beta and, to a lesser extent, by other apoptotic agents, such as TNF-a and Fas ligand.
Specifications
Specifications
| Gene ID (Entrez) | 5414 |
| Espèces | Human |
| Méthode de purification | purified |
| Numéro d’adhésion | O43236 |
| Nom usuel | Human ARTS |
| Concentration | 200 μg/mL |
| Contenu et stockage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formule | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?